The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145175.2 | NP_660158.2 | 2 - 2 | 2572 | 105 |
GenBank mRNA | AJ417080.1 | - | 2 - 2 | 2526 | 105 |