Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C7orf62
Entrez Gene ID
219557
Gene Name
chromosome 7 open reading frame 62
Gene Aliases
-
Chromosome Location
Chr.7: 88794106 - 88797937 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152706.3 NP_689919.1 2 - 2 931 141
XM_011515904.2 XP_011514206.1 2 - 2 3148 141
GenBank mRNA BC028365.2 - 2 - 2 931 141

View Full Product Details