Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MRGPRG-AS1
- Entrez Gene ID
-
283303
- Gene Name
- MRGPRG antisense RNA 1
- Gene Aliases
-
C11orf36, HSD-40
- Chromosome Location
-
Chr.11: 3218332 - 3223131 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027138.1 |
|
2 - 2 |
1320 |
104 |
GenBank mRNA |
AK093421.1 |
- |
2 - 2 |
1320 |
104 |
AK097749.1 |
- |
3 - 3 |
1353 |
104 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB337414.1 |
- |
1 |
|
|
Chr. 11 - 3222580 - 3223131 [-] on Build GRCh38 |
104 |
GenBankmRNA |
DB338484.1 |
- |
1 |
|
|
Chr. 11 - 3222565 - 3223126 [-] on Build GRCh38 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
MAS related GPR family member G |
MRGPRG |
GPR169, MRGG |
-2.8kb
|
Anti-sense |
Chr. - 3217944 - 3218813 on Build GRCh38 |
Downstream |
MAS related GPR family member E |
MRGPRE |
GPR167, MGRF, MRGE |
4.7kb
|
Anti-sense |
Chr. - 3227811 - 3232386 on Build GRCh38 |
View Full Product Details