The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002564.3 | NP_002555.3 | 3 - 3 | 2337 | 147 |
NM_176071.2 | NP_788085.2 | 3 - 3 | 2402 | 147 | |
NM_176072.2 | NP_788086.2 | 3 - 3 | 2471 | 147 | |
XM_005274019.4 | XP_005274076.1 | 3 - 3 | 2289 | 147 | |
XM_005274020.4 | XP_005274077.1 | 3 - 3 | 3876 | 147 | |
XM_005274021.4 | XP_005274078.1 | 3 - 3 | 3470 | 147 | |
XM_011545074.2 | XP_011543376.1 | 3 - 3 | 2224 | 147 | |
XM_017017839.1 | XP_016873328.1 | 4 - 4 | 2349 | 147 | |
GenBank mRNA | BC012104.1 | - | 3 - 3 | 2470 | 147 |