Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR6B1
Entrez Gene ID
135946
Gene Name
olfactory receptor family 6 subfamily B member 1
Gene Aliases
OR7-3, OR7-9
Chromosome Location
Chr.7: 144003997 - 144004932 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005281.1 NP_001005281.1 1 - 1 656 82
GenBank mRNA BC137042.1 - 1 - 1 724 82
BC137047.1 - 1 - 1 724 82

View Full Product Details