Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FHOD1 | NM_001318202.1 | NP_001305131.1 | ||
NM_013241.2 | NP_037373.2 | |||
XM_011523043.2 | XP_011521345.1 | |||
XM_011523044.1 | XP_011521346.1 | |||
XM_011523045.2 | XP_011521347.1 | |||
AB041046.1 | BAD06250.1 | |||
AB209584.1 | BAD92821.1 | |||
AF113615.1 | AAD39906.1 | |||
AK301157.1 | ||||
AK304232.1 | ||||
AY192154.1 | AAO38757.1 | |||
BC033084.1 | AAH33084.1 | |||
KJ658417.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr16:67153308 - 68071571 on Build GRCh38 | Gain | C16orf86 DUS2 DDX28 TMEM208 PLEKHG4 ACD DPEP2 ATP6V0D1 DPEP3 SLC12A4 FAM65A TSNAXIP1 CARMIL2 ELMO3 ENKD1 FHOD1 MIR328 HSD11B2 THAP11 GFOD2 CENPT NOL3 SLC9A5 NRN1L LCAT CTCF EXOC3L1 LOC100131303 LOC107984813 LRRC36 PARD6A LOC105369155 TRADD HSF4 FBXL8 CTRL TPPP3 ZDHHC1 LRRC29 EDC4 PSMB10 LOC100505942 PSKH1 AGRP E2F4 KCTD19 RANBP10 NUTF2 KIAA0895L |
nsv827707 | Chr16:67148355 - 67289296 on Build GRCh38 | Loss | SLC9A5 LRRC29 TMEM208 EXOC3L1 C16orf70 PLEKHG4 LOC105369155 TRADD HSF4 ELMO3 FBXL8 FHOD1 MIR328 B3GNT9 E2F4 KIAA0895L NOL3 |
nsv952044 | Chr16:67141198 - 67244397 on Build GRCh38 | Deletion | LRRC29 TMEM208 EXOC3L1 C16orf70 LOC105369155 TRADD HSF4 ELMO3 FBXL8 FHOD1 MIR328 B3GNT9 E2F4 KIAA0895L NOL3 |
nsv457513 | Chr16:67181417 - 67282697 on Build GRCh38 | Loss | SLC9A5 LRRC29 ELMO3 FHOD1 TMEM208 MIR328 EXOC3L1 E2F4 PLEKHG4 KIAA0895L LOC105369155 |
nsv524492 | Chr16:67174030 - 67294550 on Build GRCh38 | Loss | SLC9A5 LRRC29 TMEM208 EXOC3L1 PLEKHG4 LOC105369155 ELMO3 FHOD1 MIR328 E2F4 KCTD19 KIAA0895L NOL3 |
nsv524363 | Chr16:67136529 - 68071571 on Build GRCh38 | Loss | C16orf86 DUS2 DDX28 TMEM208 C16orf70 PLEKHG4 ACD DPEP2 ATP6V0D1 DPEP3 SLC12A4 FAM65A TSNAXIP1 CARMIL2 ELMO3 ENKD1 FHOD1 MIR328 HSD11B2 THAP11 GFOD2 CENPT NOL3 SLC9A5 NRN1L LCAT CTCF EXOC3L1 LOC100131303 LOC107984813 LRRC36 PARD6A LOC105369155 TRADD HSF4 FBXL8 CTRL TPPP3 B3GNT9 ZDHHC1 LRRC29 EDC4 PSMB10 LOC100505942 PSKH1 AGRP E2F4 KCTD19 RANBP10 NUTF2 KIAA0895L |