Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FOXP3 | NM_001114377.1 | 11 | 1320 | NP_001107849.1 |
NM_014009.3 | 12 | 1425 | NP_054728.2 | |
XM_006724533.2 | 11 | 1600 | XP_006724596.2 | |
XM_011543916.2 | 11 | 3030 | XP_011542218.1 | |
XM_017029565.1 | 13 | 2886 | XP_016885054.1 | |
XM_017029566.1 | 12 | 2850 | XP_016885055.1 | |
XM_017029567.1 | 10 | 1425 | XP_016885056.1 | |
AF277993.1 | 12 | 1425 | AAG53607.1 | |
AJ005891.1 | 10 | 1267 | CAA06748.1 | |
AK292052.1 | 12 | 1309 | ||
AK299988.1 | 11 | 1204 | ||
BC113401.1 | 11 | 1267 | ||
BC113403.1 | 11 | 1267 | ||
BC143785.1 | 10 | 1162 | ||
BC143786.1 | 10 | 1336 | ||
DQ010327.1 | 10 | 1132 | AAY27088.1 | |
EF534714.1 | 11 | 1237 | ||
EU855812.1 | 10 | 1156 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | ChrX:49205142 - 49320261 on Build GRCh38 | Gain | CCDC22 PPP1R3F FOXP3 GAGE10 CACNA1F |
nsv528292 | ChrX:48433286 - 49274368 on Build GRCh38 | Gain | SLC38A5 MAGIX PIM2 HDAC6 WDR45 ERAS SYP-AS1 SUV39H1 EBP CCDC120 RBM3 TBC1D25 GRIPAP1 PRICKLE3 TIMM17B CCDC22 PCSK1N OTUD5 GATA1 PPP1R3F FOXP3 SLC35A2 SYP CACNA1F TFE3 PLP2 PORCN KCND1 PRAF2 WAS PQBP1 GPKOW WDR13 GLOD5 FTSJ1 LOC101927635 |
esv32909 | ChrX:48003374 - 49285646 on Build GRCh38 | Gain+Loss | SSX4B ZNF182 SLC38A5 MAGIX PIM2 HDAC6 WDR45 ERAS SYP-AS1 SSX6 SUV39H1 EBP CCDC120 SSX5 ZNF630-AS1 SSX9 RBM3 TBC1D25 GRIPAP1 PRICKLE3 TIMM17B SPACA5B CCDC22 PCSK1N OTUD5 GATA1 PPP1R3F SSX4 FOXP3 SLC35A2 SYP ZNF630 CACNA1F TFE3 PLP2 PORCN KCND1 PRAF2 WAS PQBP1 GPKOW WDR13 GLOD5 SPACA5 FTSJ1 SSX1 LOC101927635 SSX3 |
nsv469734 | ChrX:49205141 - 49320261 on Build GRCh38 | Gain+Loss | CCDC22 PPP1R3F FOXP3 GAGE10 CACNA1F |
nsv6895 | ChrX:49227319 - 49262690 on Build GRCh38 | Insertion | CCDC22 FOXP3 CACNA1F |