Product Details

Assay Reference Genome
Location

Chr.15:42736060 on build GRCh38
Cytoband
15q15.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
146059
Gene Symbol
CDAN1
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
Location
Chr.15:42723557-42737219 on build GRCh38
Assay Gene Location
Overlaps Intron 2 - Exon 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CDAN1 NM_138477.2 NP_612486.2
XM_005254176.4 XP_005254233.1
XM_011521270.2 XP_011519572.1
XM_011521271.2 XP_011519573.1
AF525398.1 AAO14994.1
BC052568.1 AAH52568.1
BC066640.1 AAH66640.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv528725 Chr15:42463357 - 42812208 on Build GRCh38 Gain ZNF106 LRRC57 HAUS2 CDAN1 TTBK2 STARD9 SNAP23
nsv952615 Chr15:42714903 - 42736802 on Build GRCh38 Deletion CDAN1 STARD9
nsv457122 Chr15:42656664 - 42770350 on Build GRCh38 Loss CDAN1 TTBK2 STARD9

View Full Product Details