Product Details

Assay Reference Genome
Location

Chr.X:41474702 on build GRCh38
Cytoband
Xp11.4
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
60506
Gene Symbol
NYX
Gene Name
nyctalopin
Gene Aliases
CLRP, CSNB1, CSNB1A, CSNB4, NBM1
Location
Chr.X:41447460-41476414 on build GRCh38
Assay Gene Location
Within Exon 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NYX NM_022567.2 2 1679 NP_072089.1
XM_017029709.1 3 1347 XP_016885198.1
AF254868.1 3 1347 AAG42685.1
AJ278865.1 2 1705 CAC19014.1
BC112242.1 2 1293 AAI12243.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv25432 ChrX:41473588 - 41474938 on Build GRCh38 Loss NYX
nsv508763 ChrX:41415959 - 41531609 on Build GRCh38 Insertion CASK NYX
esv33720 ChrX:40891487 - 43157518 on Build GRCh38 Loss GPR82 LOC643043 LOC105373185 CASK LOC107985678 NYX DDX3X USP9X PPP1R2P9 GPR34

View Full Product Details