The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015916.4 | NP_057000.2 | 3 - 3 | 941 | 76 |
NR_024552.1 | 3 - 3 | 838 | 76 | ||
NR_046344.1 | 3 - 3 | 838 | 76 | ||
XM_006717880.2 | XP_006717943.1 | 3 - 3 | 817 | 76 | |
XM_006717883.2 | XP_006717946.1 | 2 - 2 | 702 | 76 | |
XM_006717884.3 | XP_006717947.1 | 3 - 3 | 1461 | 76 | |
XM_011539848.2 | XP_011538150.1 | 3 - 3 | 1473 | 76 | |
XM_017016306.1 | XP_016871795.1 | 3 - 3 | 1400 | 76 | |
XM_017016307.1 | XP_016871796.1 | 3 - 3 | 1307 | 76 | |
XM_017016308.1 | XP_016871797.1 | 3 - 3 | 955 | 76 | |
XM_017016309.1 | XP_016871798.1 | 2 - 2 | 702 | 76 | |
GenBank mRNA | AF131810.1 | - | 3 - 3 | 699 | 76 |
AK022195.1 | - | 3 - 3 | 836 | 76 | |
AK125276.1 | - | 3 - 3 | 1528 | 76 | |
BC000039.2 | - | 3 - 3 | 854 | 76 |