Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HLA-DRB3
Entrez Gene ID
3125
Gene Name
major histocompatibility complex, class II, DR beta 3
Gene Aliases
DRB1, DRw10, HLA-DR1B, HLA-DR3B, HLA-DRB, SS1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022555.3 NP_072049.2 1 - 2 139 74
GenBank mRNA AF199236.2 - 1 - 2 120 74
BC008987.1 - 1 - 2 123 74
U66825.1 - 1 - 2 139 74
U95819.1 - 1 - 2 113 74
U95820.1 - 1 - 2 43 74

View Full Product Details