The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005512.2 | NP_001005512.2 | 1 - 1 | 717 | 145 |
GenBank mRNA | BC140738.1 | - | 1 - 1 | 793 | 145 |