The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005566.2 | NP_001005566.1 | 1 - 1 | 207 | 134 |
GenBank mRNA | BC136922.1 | - | 1 - 1 | 210 | 134 |
BC136923.1 | - | 1 - 1 | 204 | 134 |