The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011717.1 | NP_001011717.1 | 6 - 7 | 346 | 151 |
GenBank mRNA | AB164464.1 | - | 6 - 7 | 346 | 151 |