Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PRR20A
- Entrez Gene ID
-
122183
- Gene Name
- proline rich 20A
- Gene Aliases
-
PRR20
- Chromosome Location
-
Chr.13: 57140918 - 57143939 on Build GRCh38
- Gene Symbol
- PRR20B
- Entrez Gene ID
-
729233
- Gene Name
- proline rich 20B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 57147488 - 57150509 on Build GRCh38
- Gene Symbol
- PRR20C
- Entrez Gene ID
-
729240
- Gene Name
- proline rich 20C
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 57154061 - 57157082 on Build GRCh38
- Gene Symbol
- PRR20D
- Entrez Gene ID
-
729246
- Gene Name
- proline rich 20D
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 57160632 - 57163653 on Build GRCh38
- Gene Symbol
- PRR20E
- Entrez Gene ID
-
729250
- Gene Name
- proline rich 20E
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 57167197 - 57170218 on Build GRCh38
View Full Product Details