The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002294.2 | NP_001002294.1 | 9 - 9 | 1867 | 126 |
NM_001319173.1 | NP_001306102.1 | 10 - 10 | 1977 | 126 | |
NM_001319174.1 | NP_001306103.1 | 8 - 8 | 1678 | 126 | |
NM_006894.5 | NP_008825.4 | 9 - 9 | 1850 | 126 | |
GenBank mRNA | AI478384.1 | - | 1 - 1 | 126 | |
BC032016.1 | - | 9 - 9 | 1802 | 126 | |
M83772.1 | - | 9 - 9 | 126 |