Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPIN4
Entrez Gene ID
139886
Gene Name
spindlin family member 4
Gene Aliases
TDRD28
Chromosome Location
Chr.X: 63347228 - 63351339 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012968.2 NP_001012986.2 1 - 1 2185 160
GenBank mRNA AK126931.1 - 1 - 1 2162 160
AL833314.1 - 1 - 1 2178 160
BC035088.1 - 1 - 1 398 160
BC043412.1 - 1 - 1 1697 160

View Full Product Details