The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144950.1 | NP_001138422.1 | 14 - 14 | 4729 | 125 |
GenBank mRNA | AF086519.1 | - | 1 - 1 | 253 | 125 |
BC130538.1 | - | 1 - 1 | 221 | 125 | |
BC130540.1 | - | 1 - 1 | 221 | 125 | |
EU699476.1 | - | 14 - 14 | 4665 | 125 |