The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024401.2 | NP_001019572.1 | 3 - 4 | 1216 | 100 |
XM_005255315.3 | XP_005255372.1 | 3 - 4 | 1250 | 100 | |
GenBank mRNA | AY874862.2 | - | 3 - 4 | 1216 | 100 |