The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003385.4 | NP_003376.2 | 2 - 3 | 553 | 82 |
GenBank mRNA | AB001104.1 | - | 2 - 3 | 308 | 82 |
AF039555.1 | - | 2 - 3 | 361 | 82 | |
BC022012.2 | - | 2 - 3 | 390 | 82 | |
BI544338.1 | - | 2 - 3 | 554 | 82 | |
U14747.1 | - | 2 - 3 | 376 | 82 |