The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009616.3 | NP_001009616.1 | 2 - 2 | 515 | 100 |
GenBank mRNA | AL039107.1 | - | 1 - 1 | 111 | 100 |