Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCB9
Entrez Gene ID
23457
Gene Name
ATP binding cassette subfamily B member 9
Gene Aliases
EST122234, TAPL
Chromosome Location
Chr.12: 122918708 - 122975160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243014.1 NP_001229943.1 8 - 8 1778 121
NM_019624.3 NP_062570.1 7 - 7 1649 121
NM_019625.3 NP_062571.1 8 - 8 1778 121
NM_203444.3 NP_982269.2 8 - 8 1778 121
XM_005253558.2 XP_005253615.1 8 - 8 1718 121
XM_011538095.2 XP_011536397.1 8 - 8 2001 121
XM_011538096.2 XP_011536398.1 8 - 8 1797 121
XM_011538098.2 XP_011536400.1 7 - 7 2643 121
XM_017019103.1 XP_016874592.1 9 - 9 1839 121
GenBank mRNA AB040953.2 - 8 - 8 4307 121
AB045381.2 - 7 - 7 1527 121
AB112582.1 - 8 - 8 1729 121
AB112583.1 - 8 - 8 1729 121
AB177852.1 - 8 - 8 1690 121
AF216494.1 - 8 - 8 1766 121
AK074738.1 - 5 - 5 722 121
AK294225.1 - 2 - 2 337 121
BC017348.2 - 7 - 7 1493 121
BC064384.1 - 8 - 8 2046 121

View Full Product Details