Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KAZN
- Entrez Gene ID
-
23254
- Gene Name
- kazrin, periplakin interacting protein
- Gene Aliases
-
KAZ
- Chromosome Location
-
Chr.1: 13893387 - 15118048 on Build GRCh38
- Gene Symbol
- TMEM51-AS1
- Entrez Gene ID
-
200197
- Gene Name
- TMEM51 antisense RNA 1
- Gene Aliases
-
C1orf126
- Chromosome Location
-
Chr.1: 15111815 - 15152464 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027136.1 |
|
5 - 5 |
|
127 |
View Full Product Details