The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207043.1 | NP_996926.1 | 4 - 4 | 1454 | 82 |
NM_207044.1 | NP_996927.1 | 3 - 3 | 1406 | 82 | |
NM_207047.1 | NP_996930.1 | 3 - 3 | 1270 | 82 | |
GenBank mRNA | AF067170.1 | - | 3 - 3 | 1364 | 82 |
AK290064.1 | - | 3 - 3 | 1253 | 82 | |
BC068544.1 | - | 3 - 3 | 1371 | 82 | |
BM771663.1 | - | 1 - 1 | 52 | 82 | |
CR749580.1 | - | 3 - 3 | 1430 | 82 |