Product Details

Assay Reference Genome
Location

Chr.7:30979074 on build GRCh38
Cytoband
7p14.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2692
Gene Symbol
GHRHR
Gene Name
growth hormone releasing hormone receptor
Gene Aliases
GHRFR, GRFR, IGHD1B
Location
Chr.7:30964021-30979531 on build GRCh38
Assay Gene Location
Within Intron 12
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GHRHR NM_000823.3 NP_000814.2
XM_011515263.1 XP_011513565.1
AB058895.1 BAB40232.1
AF282259.1 AAG15368.1
AF282260.1 AAG15369.1
AF282261.1 AAG15370.1
AF282262.1 AAG15371.1
AY557192.1 AAS59864.1
DB348314.1
L01406.1 AAA35890.1
L09237.1 AAA58619.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv6316n100 Chr7:30609596 - 31145482 on Build GRCh38 Gain INMT-FAM188B FAM188B ADCYAP1R1 AQP1 GHRHR INMT CRHR2 GARS
nsv527038 Chr7:30617851 - 31122331 on Build GRCh38 Gain INMT-FAM188B FAM188B ADCYAP1R1 AQP1 GHRHR INMT CRHR2 GARS
esv2761316 Chr7:30615466 - 31127062 on Build GRCh38 Gain INMT-FAM188B FAM188B ADCYAP1R1 AQP1 GHRHR INMT CRHR2 GARS
nsv436557 Chr7:30288062 - 32730902 on Build GRCh38 Deletion GGCT CCDC129 AVL9 NOD1 AQP1 LINC01176 LOC101928268 ADCYAP1R1 ZNRF2P1 GHRHR MIR550B1 LOC100130673 INMT CRHR2 NEUROD6 GARS ZNRF2 INMT-FAM188B DPY19L1P1 FAM188B PPP1R17 PDE1C LOC401320 MIR550A1 DKFZP586I1420 LSM5

View Full Product Details