The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199633.1 | NP_001186562.1 | 18 - 18 | 2200 | 70 |
NM_022127.2 | NP_071410.1 | 19 - 19 | 2146 | 70 | |
NR_037638.2 | 19 - 19 | 2376 | 70 | ||
XM_011518905.2 | XP_011517207.1 | 18 - 18 | 2270 | 70 | |
XM_011518906.2 | XP_011517208.1 | 19 - 19 | 2366 | 70 | |
XM_011518907.2 | XP_011517209.1 | 17 - 17 | 2085 | 70 | |
XM_011518908.2 | XP_011517210.1 | 13 - 13 | 2105 | 70 | |
GenBank mRNA | AF305210.1 | - | 19 - 19 | 2146 | 70 |
AK292849.1 | - | 18 - 18 | 2199 | 70 | |
AK304406.1 | - | 19 - 19 | 2356 | 70 | |
BC093821.1 | - | 18 - 18 | 2083 | 70 | |
BC093823.1 | - | 18 - 18 | 2083 | 70 |