Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNMT1
Entrez Gene ID
1786
Gene Name
DNA (cytosine-5-)-methyltransferase 1
Gene Aliases
ADCADN, AIM, CXXC9, DNMT, HSN1E, MCMT, m.HsaI
Chromosome Location
Chr.19: 10133344 - 10195135 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130823.2 NP_001124295.1 41 - 41 5270 92
NM_001318730.1 NP_001305659.1 40 - 40 5150 92
NM_001318731.1 NP_001305660.1 41 - 41 5149 92
NM_001379.3 NP_001370.1 40 - 40 5222 92
GenBank mRNA AB209413.2 - 40 - 40 4957 92
AF180682.1 - 38 - 38 4729 92
AK122759.1 - 13 - 13 2906 92
BC092517.1 - 41 - 41 5029 92
HY245803.1 - 1 - 1 92
X63692.1 - 40 - 40 5223 92

View Full Product Details