The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202544.2 | NP_001189473.1 | 22 - 22 | 2914 | 133 |
NM_001202545.2 | NP_001189474.1 | 22 - 22 | 2824 | 133 | |
NM_001202546.2 | NP_001189475.1 | 22 - 22 | 2741 | 133 | |
NM_001913.4 | NP_001904.2 | 23 - 23 | 2962 | 133 | |
NM_181500.3 | NP_852477.1 | 23 - 23 | 2956 | 133 | |
XM_005250154.3 | XP_005250211.1 | 23 - 23 | 3218 | 133 | |
XM_006715855.1 | XP_006715918.1 | 23 - 23 | 3212 | 133 | |
GenBank mRNA | AB075522.1 | - | 20 - 20 | 2614 | 133 |
AK122726.1 | - | 22 - 22 | 2707 | 133 | |
AK125097.1 | - | 22 - 22 | 2809 | 133 | |
BC025422.2 | - | 23 - 23 | 2837 | 133 |