The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004562.2 | NP_004553.2 | 12 - 12 | 2796 | 123 |
NM_013987.2 | NP_054642.2 | 11 - 11 | 2712 | 123 | |
NM_013988.2 | NP_054643.2 | 9 - 9 | 2349 | 123 | |
XM_011535863.1 | XP_011534165.1 | 12 - 12 | 2793 | 123 | |
XM_017010908.1 | XP_016866397.1 | 12 - 12 | 3710 | 123 | |
XM_017010909.1 | XP_016866398.1 | 11 - 11 | 2590 | 123 | |
GenBank mRNA | AB009973.1 | - | 12 - 12 | 2764 | 123 |