The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019601.3 | NP_062547.1 | 15 - 15 | 2954 | 119 |
GenBank mRNA | AK126105.1 | - | 11 - 11 | 4709 | 119 |
BC033107.1 | - | 15 - 15 | 2954 | 119 | |
BI822588.1 | - | - | 110 | 119 |