The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020897.2 | NP_065948.1 | 8 - 8 | 3407 | 168 |
XM_011509816.2 | XP_011508118.1 | 8 - 8 | 3925 | 168 | |
XM_011509817.2 | XP_011508119.1 | 7 - 7 | 3245 | 168 | |
XM_011509818.2 | XP_011508120.1 | 7 - 7 | 2637 | 168 | |
XM_017001918.1 | XP_016857407.1 | 7 - 7 | 2619 | 168 | |
GenBank mRNA | AB040968.2 | - | 8 - 8 | 3056 | 168 |
BC000066.1 | - | 7 - 7 | 2635 | 168 | |
BC028024.1 | - | 5 - 5 | 2272 | 168 |