The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128432.2 | NP_001121904.1 | 3 - 3 | 860 | 144 |
NM_001277225.1 | NP_001264154.1 | 5 - 5 | 1683 | 144 | |
NM_020973.4 | NP_066024.1 | 5 - 5 | 1781 | 144 | |
NR_102355.1 | 5 - 5 | 1781 | 144 | ||
NR_102356.1 | 3 - 3 | 860 | 144 | ||
NR_102357.1 | 5 - 5 | 1683 | 144 | ||
GenBank mRNA | AF323990.1 | - | 5 - 5 | 1729 | 144 |
AK298377.1 | - | 5 - 5 | 1684 | 144 | |
BC029362.1 | - | 3 - 3 | 851 | 144 | |
BC070188.1 | - | 3 - 3 | 841 | 144 | |
BC109377.1 | - | 3 - 3 | 841 | 144 |