The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021013.3 | NP_066293.2 | 7 - 7 | 1663 | 98 |
XM_011524793.1 | XP_011523095.1 | 8 - 8 | 1742 | 98 | |
GenBank mRNA | BC033252.1 | - | 5 - 5 | 1000 | 98 |
BC041070.1 | - | 7 - 7 | 1682 | 98 |