Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATP8B4
- Entrez Gene ID
-
79895
- Gene Name
- ATPase phospholipid transporting 8B4 (putative)
- Gene Aliases
-
ATPIM
- Chromosome Location
-
Chr.15: 49858238 - 50182096 on Build GRCh38
View Full Product Details