The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080764.3 | NP_542942.2 | 4 - 4 | 5475 | 104 |
NR_130642.1 | 5 - 5 | 4593 | 104 | ||
NR_130643.1 | 3 - 3 | 1926 | 104 | ||
XM_011529893.2 | XP_011528195.1 | 5 - 5 | 5514 | 104 | |
XM_011529895.2 | XP_011528197.1 | 3 - 3 | 5386 | 104 | |
XM_011529896.2 | XP_011528198.1 | 3 - 3 | 5108 | 104 | |
GenBank mRNA | BC042370.1 | - | 1 - 1 | 684 | 104 |