The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182627.1 | NP_872433.1 | 4 - 4 | 1162 | 78 |
GenBank mRNA | AK314105.1 | - | 4 - 4 | 1171 | 78 |
BC054030.1 | - | 4 - 4 | 1162 | 78 | |
EF011620.1 | - | 4 - 4 | 1162 | 78 |