Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH2D4B
Entrez Gene ID
387694
Gene Name
SH2 domain containing 4B
Gene Aliases
-
Chromosome Location
Chr.10: 80537902 - 80646560 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145719.1 NP_001139191.1 7 - 7 3336 127
NM_207372.2 NP_997255.2 7 - 7 3832 127
XM_017016236.1 XP_016871725.1 7 - 7 6250 127
GenBank mRNA AK091518.1 - 2 - 2 2477 127
AK123885.1 - 7 - 7 3832 127

View Full Product Details