The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145373.2 | NP_001138845.1 | 1 - 1 | 3005 | 94 |
GenBank mRNA | AK096389.1 | - | 1 - 1 | 1850 | 94 |