Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNG10
Entrez Gene ID
2790
Gene Name
G protein subunit gamma 10
Gene Aliases
-
Chromosome Location
Chr.9: 111661571 - 111670246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017998.3 NP_001017998.1 1 - 2 146 102
NM_001198664.1 NP_001185593.1 1 - 2 146 102
GenBank mRNA AF493877.1 - 1 - 2 82 102
BC010384.1 - 1 - 2 108 102
BC015206.2 - 1 - 2 92 102
BC016319.1 - 1 - 2 104 102
BC072671.1 - 1 - 2 97 102
BC107574.1 - 1 - 2 113 102
U31383.1 - 1 - 2 105 102

View Full Product Details