Product Details

Assay Reference Genome
Location

Chr.17:46039931 on build GRCh38
Cytoband
17q21.31
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
284058
Gene Symbol
KANSL1
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
Location
Chr.17:46029916-46225374 on build GRCh38
Assay Gene Location
Overlaps Intron 12 - Exon 13
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
KANSL1 NM_001193465.1 NP_001180394.1
NM_001193466.1 NP_001180395.1
NM_015443.3 NP_056258.1
XM_006721823.1 XP_006721886.1
XM_006721824.3 XP_006721887.1
XM_011524628.2 XP_011522930.1
XM_011524629.2 XP_011522931.1
XM_011524630.2 XP_011522932.1
XM_011524631.2 XP_011522933.1
XM_011524632.2 XP_011522934.1
XM_017024488.1 XP_016879977.1
XM_017024489.1 XP_016879978.1
AB033093.1 BAA86581.1
AK094946.1 1 135
AK291259.1
AL137317.1 CAB70694.1
BC035892.1 AAH35892.1
BC098376.1 AAH98376.1
BX538006.1 CAD97958.1
BX648760.1 1 6194 CAH10565.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1072740 Chr17:45970433 - 46248034 on Build GRCh38 Deletion MAPT KANSL1 STH KANSL1-AS1
esv2751686 Chr17:45962198 - 46286690 on Build GRCh38 Gain MAPT ARL17B KANSL1 STH LRRC37A KANSL1-AS1
nsv1063349 Chr17:46039494 - 46358076 on Build GRCh38 Gain LOC107987254 ARL17B KANSL1 LRRC37A KANSL1-AS1
esv2758692 Chr17:46006548 - 47199967 on Build GRCh38 Gain+Loss ARL17A MAPT KANSL1 WNT9B NSF RPS2P47 RPRML MIR5089 CDC27 LOC107987254 WNT3 ARL17B LRRC37A LOC107987253 GOSR2 NSFP1 KANSL1-AS1 LRRC37A2
dgv128e55 Chr17:46017105 - 46286690 on Build GRCh38 Gain MAPT ARL17B KANSL1 LRRC37A KANSL1-AS1
nsv471698 Chr17:46002113 - 46300887 on Build GRCh38 Gain+Loss MAPT ARL17B KANSL1 LRRC37A KANSL1-AS1
nsv469705 Chr17:45969304 - 46127121 on Build GRCh38 Loss MAPT KANSL1 STH
nsv469537 Chr17:46002112 - 46163353 on Build GRCh38 Gain+Loss MAPT KANSL1
nsv2069 Chr17:46034385 - 46065659 on Build GRCh38 Insertion KANSL1
nsv575142 Chr17:46003698 - 46172742 on Build GRCh38 Loss MAPT KANSL1
esv2762433 Chr17:45496768 - 46710828 on Build GRCh38 Gain+Loss ARL17A LOC644172 MAPT MGC57346 KANSL1 NSF MAPT-AS1 STH CRHR1 MAPT-IT1 MGC57346-CRHR1 SPPL2C LOC107987254 ARL17B LRRC37A4P LRRC37A LOC107987253 NSFP1 KANSL1-AS1 LOC105376839 LRRC37A2 CRHR1-IT1
dgv366n27 Chr17:46032477 - 46215654 on Build GRCh38 Gain KANSL1 KANSL1-AS1
dgv5618n54 Chr17:46032108 - 46330700 on Build GRCh38 Gain ARL17B KANSL1 LRRC37A KANSL1-AS1
dgv127e55 Chr17:45927606 - 46733750 on Build GRCh38 Gain LOC107987254 ARL17A MAPT ARL17B KANSL1 NSF STH LRRC37A LOC107987253 NSFP1 KANSL1-AS1 LRRC37A2

View Full Product Details