Product Details
- Assay Reference Genome
Location
Chr.8:38411731 on build GRCh38
- Cytoband
- 8p11.23
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 2260
- Gene Symbol
- FGFR1
- Gene Name
- fibroblast growth factor receptor 1
- Gene Aliases
- BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
- Location
- Chr.8:38411138-38468834 on build GRCh38
- Assay Gene Location
- Within Exon 23
View Full Product Details