Product Details

Assay Reference Genome
Location

Chr.16:13926707 on build GRCh38
Cytoband
16p13.12
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2072
Gene Symbol
ERCC4
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Gene Aliases
ERCC11, FANCQ, RAD1, XFEPS, XPF
Location
Chr.16:13920138-13952348 on build GRCh38
Assay Gene Location
Within Exon 4
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ERCC4 NM_005236.2 3 544 NP_005227.1
XM_011522424.2 4 701 XP_011520726.1
XM_017023043.1 3 510 XP_016878532.1
AK289726.1 3 543
AK308341.1 3 510
BC020741.1 3 515
BC142631.1 3 544
U64315.1 3 517 AAB07689.1

View Full Product Details