Product Details

Assay Reference Genome
Location

Chr.3:14134807 on build GRCh38
Cytoband
3p25.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
79188
Gene Symbol
TMEM43
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
Location
Chr.3:14124940-14143680 on build GRCh38
Assay Gene Location
Overlaps Intron 8 - Exon 9
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
TMEM43 NM_024334.2 NP_077310.1
XM_017007176.1 XP_016862665.1
AK027466.1 BAB55131.1
AK027757.1 BAB55348.1
AK027827.1 BAB55396.1
AK027877.1 BAB55425.1
AK074073.1 BAB84899.1
AK075010.1 BAC11350.1
AK225589.1
AL136916.1 CAB66850.1
AY358625.1 AAQ88988.1
BC003125.1 AAH03125.1
BC008054.2 AAH08054.2
BC011719.2 AAH11719.1
KU178769.1
KU178770.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1011766 Chr3:13485961 - 14213016 on Build GRCh38 Gain SNORA93 XPC TPRXL CHCHD4 FGD5P1 LINC00620 LSM3 LOC100293612 FBLN2 HDAC11 TMEM43 WNT7A
esv34163 Chr3:13865660 - 14212447 on Build GRCh38 Loss XPC TPRXL CHCHD4 FGD5P1 LSM3 TMEM43 WNT7A
esv2724945 Chr3:13931758 - 14820330 on Build GRCh38 Deletion XPC GRIP2 CHCHD4 CCDC174 FGD5P1 LINC01267 LSM3 LOC152274 SLC6A6 C3orf20 TMEM43 TPRXL FGD5
nsv955318 Chr3:14126901 - 14142100 on Build GRCh38 Deletion TMEM43

View Full Product Details