The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178516.3 | NP_848611.2 | 9 - 10 | 1734 | 54 |
GenBank mRNA | AK092858.1 | - | 9 - 10 | 1626 | 54 |
AK092906.1 | - | 8 - 9 | 1372 | 54 | |
AK292004.1 | - | 9 - 10 | 1643 | 54 | |
AK300915.1 | - | 8 - 9 | 1393 | 54 | |
BC034287.1 | - | 6 - 7 | 1083 | 54 |