The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017521.2 | NP_059991.1 | 3 - 3 | 1128 | 73 |
GenBank mRNA | BC023511.2 | - | 3 - 3 | 1023 | 73 |
Y08976.1 | - | 3 - 3 | 1130 | 73 |