Product Details

Assay Reference Genome
Location

Chr.18:63656966 on build GRCh38
Cytoband
18q21.33
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
6317
Gene Symbol
SERPINB3
Gene Name
serpin family B member 3
Gene Aliases
HsT1196, SCC, SCCA-1, SCCA-PD, SCCA1, SSCA1, T4-A
Location
Chr.18:63655197-63661963 on build GRCh38
Assay Gene Location
Overlaps Intron 6 - Exon 7
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SERPINB3 NM_006919.2 NP_008850.1
AB046399.1 BAB40772.1
AF428135.1 AAQ04218.1
AF428136.1 AAQ04219.1
AF428137.1 AAQ04220.1
AF428138.1 AAQ04221.1
AF428139.1 AAQ04222.1
AJ515706.1 CAD56658.1
AK222746.1 BAD96466.1
AK222753.1 BAD96473.1
AK307261.1
AK314805.1
AY190327.1 AAO11731.1
AY245778.1 AAO92269.1
AY245781.1 AAO92272.1
BC005224.1 AAH05224.1
BT006748.1 AAP35394.1
EU852041.1
S66896.1 AAB20405.1
U19556.1 AAA97552.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3642778 Chr18:63643736 - 63659432 on Build GRCh38 Loss SERPINB3 SERPINB4
nsv833672 Chr18:63620445 - 63772481 on Build GRCh38 Gain SERPINB3 SERPINB4 SERPINB7 SERPINB11
nsv1066552 Chr18:63147299 - 64608176 on Build GRCh38 Gain SERPINB3 SERPINB8 LINC01538 SERPINB4 LINC00305 SERPINB7 KDSR SERPINB11 VPS4B SERPINB12 SERPINB13 SERPINB5 SERPINB2 BCL2 HMSD LOC284294 SERPINB10
nsv1057179 Chr18:63646569 - 63679645 on Build GRCh38 Loss SERPINB3
esv2717205 Chr18:63641023 - 63662901 on Build GRCh38 Deletion SERPINB3 SERPINB4

View Full Product Details