Product Details

Assay Reference Genome
Location

Chr.15:34343039 on build GRCh38
Cytoband
15q14
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55505
Gene Symbol
NOP10
Gene Name
NOP10 ribonucleoprotein
Gene Aliases
DKCB1, NOLA3, NOP10P
Location
Chr.15:34341716-34343161 on build GRCh38
Assay Gene Location
Overlaps Intron 1 - Exon 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NOP10 NM_018648.3 NP_061118.1
AB043104.1 BAA96133.1
BC008886.2 AAH08886.1
BC063023.1 AAH63023.1
BM554889.1
BQ218536.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv471682 Chr15:34268250 - 34590429 on Build GRCh38 Gain+Loss NOP10 MIR1233-1 MIR1233-2 GOLGA8A NUTM1 SLC12A6 GOLGA8B LPCAT4
nsv1052620 Chr15:32612402 - 34538731 on Build GRCh38 Gain LOC105370757 FMN1 ARHGAP11A PGBD4 EMC4 GOLGA8A AVEN NUTM1 KATNBL1 GOLGA8B NOP10 GREM1 LOC101928134 CHRM5 MIR1233-1 MIR1233-2 RYR3 SLC12A6 SCG5 TMCO5B LOC100131315 LPCAT4 LOC101928042 EMC7
esv33337 Chr15:31689863 - 34575097 on Build GRCh38 Gain+Loss GOLGA8K FMN1 PGBD4 GOLGA8A LOC100996255 NUTM1 ULK4P1 KATNBL1 GOLGA8B NOP10 GREM1 MIR1233-2 WHAMMP1 RYR3 GOLGA8O LPCAT4 LOC105370757 GOLGA8N ARHGAP11A EMC4 LOC105370940 AVEN LOC101060588 LOC101928134 LOC100653133 CHRM5 MIR1233-1 CHRNA7 SLC12A6 SCG5 TMCO5B LOC100131315 LOC101928042 EMC7
esv2760024 Chr15:34250820 - 34696046 on Build GRCh38 Gain+Loss NOP10 MIR1233-1 MIR1233-2 GOLGA8A NUTM1 SLC12A6 GOLGA8B LPCAT4
esv3569431 Chr15:33564570 - 35080307 on Build GRCh38 Gain PGBD4 EMC4 GOLGA8A AVEN NUTM1 KATNBL1 GOLGA8B AQR LOC101928174 NOP10 CHRM5 MIR1233-1 ACTC1 MIR1233-2 GJD2 RYR3 ZNF770 SLC12A6 LPCAT4 EMC7
nsv469547 Chr15:34302356 - 34465606 on Build GRCh38 Loss NOP10 MIR1233-1 GOLGA8A NUTM1 SLC12A6 LPCAT4
dgv4456n54 Chr15:32604149 - 34557061 on Build GRCh38 Gain FMN1 PGBD4 GOLGA8A NUTM1 KATNBL1 GOLGA8B NOP10 GREM1 MIR1233-2 RYR3 LPCAT4 LOC105370757 GOLGA8N ARHGAP11A EMC4 AVEN LOC101928134 CHRM5 MIR1233-1 SLC12A6 SCG5 TMCO5B LOC100131315 LOC101928042 EMC7

View Full Product Details