Product Details

Assay Reference Genome
Location

Chr.7:100417140 on build GRCh38
Cytoband
7q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55063
Gene Symbol
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Location
Chr.7:100400826-100429239 on build GRCh38
Assay Gene Location
Within Exon 7
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ZCWPW1 NM_001258008.1 6 782 NP_001244937.1
NM_017984.4 6 782 NP_060454.3
XM_005250479.2 6 1213 XP_005250536.1
XM_005250480.2 6 1213 XP_005250537.1
XM_006716035.3 6 1213 XP_006716098.1
XM_006716036.2 6 717 XP_006716099.1
XM_006716037.3 6 1213 XP_006716100.1
XM_006716038.3 6 1213 XP_006716101.1
XM_006716040.3 6 1213 XP_006716103.1
XM_017012379.1 6 1213 XP_016867868.1
XM_017012380.1 6 717 XP_016867869.1
AK000919.1 1 166 BAA91424.1
AK093038.1 6 717 BAC04028.1
AK300745.1 6 638
AK302108.1
AL136735.1 6 653 CAB66669.1
BC002725.1 3 480 AAH02725.1
KU178677.1 1 42
KU178678.1 1 42
KU178679.1 1 42
KU178680.1 1 42
KU178681.1 1 42
KU178682.1 1 42

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2734888 Chr7:99975623 - 101290285 on Build GRCh38 Deletion MIR6840 PILRB LOC102724094 PILRA ZASP PPP1R35 ZCWPW1 TSC22D4 NYAP1 ZNHIT1 C7orf61 STAG3L5P-PVRIG2P-PILRB MUC17 COPS6 PCOLCE-AS1 ZKSCAN1 UFSP1 SPDYE3 VGF LRCH4 GAL3ST4 MOSPD3 NAT16 LOC105375429 TFR2 SLC12A9 SAP25 MIR93 MUC12 CLDN15 ZNF3 C7orf43 SERPINE1 TRIP6 ZAN EPHB4 MIR4653 GIGYF1 STAG3 MCM7 MIR25 GATS PVRIG CNPY4 AZGP1P1 MEPCE EPO STAG3L5P MIR106B PMS2P1 LAMTOR4 ACTL6B ACHE MIR4658 TRIM56 PVRIG2P FIS1 ZSCAN21 MUC3A TAF6 AP1S1 AP4M1 MIR6875 GPC2 PLOD3 GNB2 MBLAC1 FBXO24 AGFG2 MOGAT3 SRRT POP7 AZGP1 PCOLCE
nsv831071 Chr7:100320402 - 100515790 on Build GRCh38 Loss MIR6840 PILRB SPDYE3 PVRIG2P PILRA PPP1R35 ZCWPW1 MEPCE STAG3L5P TSC22D4 NYAP1 C7orf61 STAG3L5P-PVRIG2P-PILRB PMS2P1
nsv527107 Chr7:100417088 - 100467096 on Build GRCh38 Loss PPP1R35 ZCWPW1 MEPCE TSC22D4 C7orf61

View Full Product Details