Product Details

Assay Reference Genome
Location

Chr.11:85958397 on build GRCh38
Cytoband
11q14.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8301
Gene Symbol
PICALM
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
Location
Chr.11:85957171-86069881 on build GRCh38
Assay Gene Location
Within Exon 23
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PICALM NM_001008660.2 20 2799 NP_001008660.1
NM_001206946.1 20 2904 NP_001193875.1
NM_001206947.1 19 2490 NP_001193876.1
NM_007166.3 20 2925 NP_009097.2
XM_005274322.2 22 3036 XP_005274379.1
XM_005274323.2 22 3021 XP_005274380.1
XM_005274324.2 21 3012 XP_005274381.1
XM_005274325.2 21 2997 XP_005274382.1
XM_005274326.2 21 2933 XP_005274383.1
XM_005274327.2 20 2952 XP_005274384.1
XM_005274328.2 21 2918 XP_005274385.1
XM_005274329.3 20 2894 XP_005274386.1
XM_005274330.2 21 2886 XP_005274387.1
XM_005274331.2 19 2849 XP_005274388.1
XM_005274332.2 21 2871 XP_005274389.1
XM_005274333.2 20 2862 XP_005274390.1
XM_005274334.2 19 2834 XP_005274391.1
XM_005274335.2 20 2847 XP_005274392.1
XM_005274336.2 20 2768 XP_005274393.1
XM_005274337.2 18 2744 XP_005274394.1
XM_005274340.2 17 2594 XP_005274397.1
XM_006718699.2 22 3015 XP_006718762.1
XM_006718700.2 22 3000 XP_006718763.1
XM_006718701.3 21 2991 XP_006718764.1
XM_011545293.1 20 2778 XP_011543595.1
XM_017018381.1 21 2897 XP_016873870.1
XM_017018382.1 19 2802 XP_016873871.1
XM_017018383.1 19 2759 XP_016873872.1
XM_017018384.1 19 2744 XP_016873873.1
XM_017018385.1 18 2729 XP_016873874.1
XM_017018386.1 18 2699 XP_016873875.1
XM_017018387.1 18 2684 XP_016873876.1
XM_017018388.1 19 2675 XP_016873877.1
AB210017.1 20 2904
AK128786.1 1 653
BC048259.1 20 2686 AAH48259.2
BC064357.1 20 2761 AAH64357.1
BC073961.1 20 2635 AAH73961.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1053808 Chr11:85877876 - 86046651 on Build GRCh38 Gain CCDC83 PICALM
nsv1035489 Chr11:85791910 - 86067187 on Build GRCh38 Gain SYTL2 CCDC83 PICALM
dgv1247n100 Chr11:85930576 - 85973892 on Build GRCh38 Gain PICALM

View Full Product Details