Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTRA3
Entrez Gene ID
94031
Gene Name
HtrA serine peptidase 3
Gene Aliases
Prsp, Tasp
Chromosome Location
Chr.4: 8269757 - 8307111 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297559.1 NP_001284488.1 5 - 6 1140 75
NM_053044.4 NP_444272.1 5 - 6 1140 75
XM_011513596.2 XP_011511898.1 5 - 6 1148 75
GenBank mRNA AY040094.2 - 5 - 6 1157 75
AY280665.1 - 5 - 6 1128 75
AY280666.1 - 5 - 6 1043 75
BC034390.1 - 5 - 6 1140 75
BC035717.1 - 5 - 6 1140 75

View Full Product Details